Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163774.1 | NP_001157246.1 | 4 | 1315 | ||
NM_001163775.1 | NP_001157247.1 | 4 | 1315 | |||
GenBank mRNA | AK031190.1 | - | 4 | 1051 | ||
AK046145.1 | - | 3 | 2550 | |||
AK129235.1 | - | 4 | 422 | |||
BC075676.1 | - | 4 | 878 | |||
BC077714.1 | - | 4 | 764 | |||
BC085152.1 | - | 4 | 1018 |