Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163774.1 | NP_001157246.1 | 15, 16 | 3120 | ||
NM_001163775.1 | NP_001157247.1 | 15, 16 | 3120 | |||
GenBank mRNA | AK129235.1 | - | 15, 16 | 2227 | ||
BC052933.1 | - | 5, 6 | 1077 | |||
BC075676.1 | - | 15, 16 | 2682 | |||
BC077714.1 | - | 15, 16 | 2569 | |||
BC085152.1 | - | 15, 16 | 2823 |