Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 6, 7 | 912 | ||
GenBank mRNA | AK173222.1 | - | 5, 6 | 1582 | ||
BC027735.1 | - | 6, 7 | 814 | |||
BC027790.1 | - | 1, 2 | 614 | |||
BC042391.1 | - | 1, 2 | 292 | |||
BU708306.1 | - | 4, 5 | 404 |