Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033471.3 | NP_001028643.2 | 3 | 1190 | ||
GenBank mRNA | AK133001.1 | - | 3 | 1192 | ||
AV270501.2 | - | 1 | 209 | |||
BC147520.1 | - | 2 | 1047 | 9056512 | ||
BC147548.1 | - | 2 | 1047 | 9056539 |