Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019177.3 | NP_062050.3 | 3 | 388 | ||
GenBank mRNA | D10831.1 | - | 3 | 380 | ||
S79523.1 | - | 3 | 388 |