Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163189.1 | NP_001156661.1 | 1 | 390 | ||
GenBank mRNA | BC158112.1 | - | 1 | 390 | 9088186 |