Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019188.1 | NP_062061.1 | 4 | 282 | ||
XM_008770989.1 | XP_008769211.1 | 4 | 1084 | |||
GenBank mRNA | U65486.1 | - | 4 | 282 |