Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099632.1 | NP_001093102.1 | 4 | 1191 | ||
GenBank mRNA | BC132405.1 | - | 4 | 1191 | 40130682 | |
BC145677.1 | - | 4 | 1191 | 40130980 |