Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019198.1 | NP_062071.1 | 5 | 580 | ||
XM_008770827.1 | XP_008769049.1 | 6 | 736 | |||
GenBank mRNA | AB008682.1 | - | 5 | 580 |