Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013799.1 | NP_001013821.1 | 3 | 295 | ||
GenBank mRNA | AK041301.1 | - | 3 | 295 | ||
AK163176.1 | - | 3 | 325 | |||
BC147328.1 | - | 3 | 233 | 9056394 | ||
BC147329.1 | - | 3 | 233 | 9056414 |