Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syndig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085521.2 | NP_001078990.1 | 4 | 1760 | ||
GenBank mRNA | BY638665.1 | - | 1 | 116 |