Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syndig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085521.2 | NP_001078990.1 | 4 | 1666 | ||
GenBank mRNA | BC147352.1 | - | 3 | 1467 | 9056389 | |
BC147353.1 | - | 3 | 1467 | 9056409 | ||
BC147660.1 | - | 3 | 1467 | 9087449 | ||
BC147661.1 | - | 3 | 1467 | 9087450 | ||
BY638665.1 | - | 1 | 22 |