Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017350.1 | NP_059046.1 | 5 | 504 | ||
NM_134352.1 | NP_599179.1 | 5 | 509 | |||
GenBank mRNA | AF007789.1 | - | 5 | 496 | ||
BC127499.1 | - | 5 | 504 | 7380245 | ||
FQ220340.1 | - | 5 | 517 | |||
X71898.1 | - | 5 | 509 | |||
X71899.1 | - | 5 | 504 |