Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081265.1 | NP_001074734.1 | 6 | 1326 | ||
GenBank mRNA | BC147492.1 | - | 6 | 1320 | 9056289 | |
BC158066.1 | - | 6 | 784 | 9007365 |