Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017358.1 | NP_059054.1 | 4 | 554 | ||
XM_006242791.2 | XP_006242853.1 | 4 | 592 | |||
XM_006242792.2 | XP_006242854.1 | 5 | 1181 | |||
XM_006242793.2 | XP_006242855.1 | 4 | 592 | |||
XM_008766066.1 | XP_008764288.1 | 5 | 1991 | |||
GenBank mRNA | AF004840.1 | - | 4 | 554 |