Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079932.3 | NP_001073401.1 | 6 | 1685 | ||
GenBank mRNA | AB231474.1 | - | 6 | 1589 | ||
BC067209.1 | - | 6 | 1251 | |||
BC147789.1 | - | 6 | 1685 | 9087507 | ||
BC158111.1 | - | 6 | 1670 | 9088185 | ||
BC158120.1 | - | 6 | 1670 | 9088192 | ||
BC172156.1 | - | 5 | 1662 |