Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017361.1 | NP_059057.1 | 3 | 252 | ||
GenBank mRNA | BC078858.1 | - | 3 | 282 | 7134682 | |
U63840.1 | - | 3 | 252 |