Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253914.2 | 4 | 519 | |||
GenBank mRNA | FQ220053.1 | - | 523 | |||
FQ220072.1 | - | 519 | ||||
FQ220274.1 | - | 521 | ||||
FQ220568.1 | - | 521 | ||||
FQ220587.1 | - | 519 | ||||
FQ220864.1 | - | 245 | ||||
FQ221231.1 | - | 521 | ||||
FQ222593.1 | - | 519 | ||||
FQ223300.1 | - | 522 | ||||
FQ228787.1 | - | 519 | ||||
FQ229328.1 | - | 519 | ||||
FQ229537.1 | - | 518 | ||||
FQ229656.1 | - | 513 | ||||
FQ230119.1 | - | 519 | ||||
M55269.1 | - | 512 |
Gene Symbol: LOC100911206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257188.2 | XP_006257250.2 | 1 | 59 |
Gene Symbol: LOC103690150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771672.1 | XP_008769894.1 | 496 | |||
GenBank mRNA | FQ228618.1 | - | 519 |
Gene Symbol: Prl3d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771671.1 | XP_008769893.1 | 4 | 522 | ||
GenBank mRNA | BC161851.1 | - | 4 | 496 | 9088452 | |
D21103.1 | - | 4 | 458 | |||
DQ329282.1 | - | 4 | 515 | |||
DQ329283.1 | - | 4 | 515 | |||
FQ145043.1 | - | 4 | 521 | |||
FQ221365.1 | - | 4 | 519 | |||
FQ223255.1 | - | 4 | 522 | |||
FQ223518.1 | - | 4 | 519 | |||
FQ229928.1 | - | 4 | 521 | |||
FQ230035.1 | - | 4 | 519 |