Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033785.2 | NP_001028957.1 | 2 | 1136 | ||
GenBank mRNA | AK133120.1 | - | 2 | 1137 | ||
BC147265.1 | - | 2 | 1136 | 9056322 | ||
BC147494.1 | - | 2 | 1136 | 9056315 |