Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034059.1 | NP_001029231.1 | 2 | 1411 | ||
GenBank mRNA | AK134708.1 | - | 2 | 1411 |