Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dupd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013826.2 | NP_001013848.1 | 2 | 283 | ||
GenBank mRNA | AK075665.1 | - | 2 | 284 | ||
BC147517.1 | - | 2 | 246 | 9056509 | ||
BC147531.1 | - | 2 | 246 | 9056522 |