Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50487
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
4
546
NM_001034069.1
NP_001029241.1
4
546
NM_001034070.1
NP_001029242.1
4
546
NM_001034071.1
NP_001029243.1
4
546
NM_001034072.1
NP_001029244.1
4
546
NM_001034073.1
NP_001029245.1
3
483
NM_001034074.1
NP_001029246.1
3
483
NM_001034075.1
NP_001029247.1
3
483
NM_001301336.1
NP_001288265.1
4
546
NM_001301342.1
NP_001288271.1
4
546
NM_001301736.1
NP_001288665.1
3
483
NM_019131.2
NP_062004.1
3
483
XM_006243386.2
XP_006243448.1
4
538
XM_006243388.2
XP_006243450.1
4
538
XM_006243389.2
XP_006243451.1
4
538
XM_006243390.2
XP_006243452.1
4
538
XM_006243393.1
XP_006243455.1
3
481
XM_006243394.1
3
499
XM_006243395.2
XP_006243457.1
3
490
XM_008766207.1
XP_008764429.1
4
538
XM_008766208.1
XP_008764430.1
3
481
GenBank mRNA
AF372215.1
-
3
332
AF372216.1
-
4
445
AF372217.1
-
4
451
BC078780.1
-
3
450
7106337
BU670983.1
-
4
546
CB585296.1
-
3
486
CF108680.1
-
4
494
CK483405.1
-
4
500
FQ074026.1
-
4
604
FQ214803.1
-
4
517
FQ214809.1
-
4
515
FQ214835.1
-
4
515
FQ214864.1
-
4
519
FQ214868.1
-
4
515
FQ214869.1
-
4
521
FQ214872.1
-
4
514
FQ214877.1
-
4
515
FQ214894.1
-
4
521
FQ214902.1
-
4
515
FQ214911.1
-
4
517
FQ214944.1
-
4
521
FQ214946.1
-
4
521
FQ214991.1
-
4
515
FQ215107.1
-
4
515
FQ215186.1
-
4
521
FQ215188.1
-
4
515
FQ215206.1
-
4
550
FQ215459.1
-
4
517
FQ215512.1
-
4
521
FQ215522.1
-
4
592
FQ215524.1
-
4
517
FQ215538.1
-
4
521
FQ215551.1
-
4
516
FQ215698.1
-
4
515
FQ215703.1
-
4
515
FQ215724.1
-
4
602
FQ215746.1
-
4
517
FQ215761.1
-
4
515
FQ215768.1
-
4
515
FQ215803.1
-
4
521
FQ215808.1
-
4
517
FQ215815.1
-
4
515
FQ215856.1
-
4
515
FQ215919.1
-
4
523
FQ215944.1
-
4
522
FQ215969.1
-
4
515
FQ215995.1
-
4
517
FQ216014.1
-
4
599
FQ216018.1
-
4
521
FQ216052.1
-
4
515
FQ216082.1
-
4
599
FQ216105.1
-
4
515
FQ216118.1
-
4
599
FQ216145.1
-
4
521
FQ216160.1
-
4
514
FQ216319.1
-
4
517
FQ216340.1
-
4
521
FQ216368.1
-
4
517
FQ216400.1
-
4
517
FQ216494.1
-
4
515
FQ216552.1
-
4
521
FQ216557.1
-
4
521
FQ216569.1
-
610
FQ216615.1
-
4
430
FQ216637.1
-
4
517
FQ216683.1
-
4
517
FQ216697.1
-
4
517
FQ216747.1
-
4
517
FQ217468.1
-
3
266
FQ217503.1
-
4
521
FQ217616.1
-
2
83
FQ217637.1
-
4
521
FQ217808.1
-
4
517
FQ223706.1
-
4
517
FQ223993.1
-
4
519
FQ224036.1
-
4
432
FQ224207.1
-
4
515
FQ224838.1
-
4
521
FQ224943.1
-
4
519
FQ224961.1
-
4
517
M34134.1
-
3
468
M34135.1
-
4
480
M34136.1
-
2
198
M60666.1
-
4
493
M60667.1
-
4
493
M60668.1
-
3
430
M60669.1
-
3
430
X02411.1
-
4
444
X02412.1
-
2
200
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline