Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038997.2 | NP_001034086.1 | 2 | 103 | ||
GenBank mRNA | BC132026.1 | - | 2 | 103 | 40130303 | |
BC132030.1 | - | 2 | 103 | 40130307 |