Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016971.2 | NP_058667.1 | 5 | 884 | ||
GenBank mRNA | AJ249491.1 | - | 5 | 910 |