Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033128.3 | NP_001028300.1 | 17 | 5221 | ||
GenBank mRNA | BC076577.1 | - | 17 | 5210 |