Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081279.1 | NP_001074748.1 | 3 | 3745 | ||
GenBank mRNA | AK081646.1 | - | 3 | 710 | ||
AK132349.1 | - | 3 | 2954 | |||
AK171731.1 | - | 3 | 2954 | |||
AK171759.1 | - | 3 | 2315 | |||
BC082308.1 | - | 3 | 3745 |