Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D13Ertd608e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143258.1 | - | 2 | 293 | ||
BC147106.1 | - | 2 | 288 | 9056131 | ||
BC147107.1 | - | 2 | 288 | 9056120 |