Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029148.1 | NP_083424.1 | 8 | 2950 | ||
GenBank mRNA | AK013100.1 | - | 1 | 134 | ||
AK129301.1 | - | 8 | 2949 | |||
AK147465.1 | - | 8 | 2951 | |||
BC050918.1 | - | 8 | 2849 | |||
BC054558.1 | - | 8 | 2950 | 6405599 |