Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039103.3 | NP_001034192.1 | 7 | 828 | ||
NM_133914.3 | NP_598675.2 | 7 | 828 | |||
GenBank mRNA | AK014220.1 | - | 7 | 747 | ||
AK044762.1 | - | 7 | 656 | |||
AK133565.1 | - | 7 | 747 | |||
AK154335.1 | - | 7 | 829 | |||
AK155146.1 | - | 7 | 829 | |||
AY591339.1 | - | 7 | 729 | |||
BC027230.1 | - | 7 | 719 | |||
BC049381.1 | - | 7 | 803 | |||
BC057460.1 | - | 7 | 682 | 4527934 | ||
DQ317932.1 | - | 7 | 712 |