Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 6 | 976 | ||
GenBank mRNA | AJ237585.1 | - | 1 | 39 | ||
AK017756.1 | - | 6 | 870 | |||
CX564513.1 | - | 5 | 621 |