Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027711.1 | NP_081987.1 | 23 | 2971 | ||
GenBank mRNA | AK042038.1 | - | 8 | 863 | ||
AK142813.1 | - | 23 | 2972 | |||
AK147360.1 | - | 23 | 2965 | |||
AK171697.1 | - | 3 | 439 | |||
AK172491.1 | - | 3 | 439 | |||
BC150958.1 | - | 23 | 2970 | 9087869 |