Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080971.2 | NP_001074440.1 | 2 | 260 | ||
GenBank mRNA | BC147322.1 | - | 2 | 158 | 9056380 | |
BC147323.1 | - | 2 | 158 | 9056384 | ||
BC147687.1 | - | 2 | 260 | 9056660 | ||
BC147698.1 | - | 2 | 260 | 9056671 |