Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019158.2 | NP_062031.1 | 4 | 538 | ||
XM_006230187.2 | XP_006230249.1 | 4 | 585 | |||
GenBank mRNA | AB005547.1 | - | 4 | 537 | ||
AF007775.1 | - | 4 | 517 | |||
BC081812.1 | - | 4 | 538 | 7114305 |