Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166260.1 | NP_001159732.1 | 2, 3 | 396 | ||
NM_004237.3 | NP_004228.1 | 2, 3 | 396 | |||
XM_011514163.2 | XP_011512465.1 | 2, 3 | 366 | |||
GenBank mRNA | AK310376.1 | - | 2, 3 | 366 | ||
BC000404.2 | - | 2, 3 | 361 | 2822981 | ||
BC019294.1 | - | 2, 3 | 361 | 2822981 | ||
CR456744.1 | - | 2, 3 | 251 | |||
L40384.1 | - | 2, 3 | 257 | |||
U96131.1 | - | 2, 3 | 296 |