Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081182.2 | NP_001074651.2 | 28 | 5127 | ||
GenBank mRNA | AK173107.1 | - | 17 | 4572 |