Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253916.1 | NP_001240845.1 | 17 | 5362 | ||
NM_001253917.1 | NP_001240846.1 | 16 | 5164 | |||
NM_019982.3 | NP_064366.2 | 17 | 5359 | |||
GenBank mRNA | AF220355.1 | - | 1 | 1933 | ||
AK042828.1 | - | 1 | 742 | |||
AK122401.1 | - | 15 | 4284 | |||
BC056471.1 | - | 12 | 3774 | |||
BC065117.1 | - | 17 | 5317 | 6840338 | ||
BC065129.1 | - | 17 | 5138 | 30362651 | ||
BC079593.1 | - | 11 | 3702 | |||
BC094376.1 | - | 11 | 3673 | |||
BC099839.1 | - | 11 | 2177 | |||
DQ167195.1 | - | 17 | 5346 |