Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn34b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033799.2 | NP_001028971.1 | 3 | 459 | ||
GenBank mRNA | AK132827.1 | - | 3 | 460 | ||
BC147626.1 | - | 3 | 329 | 9056616 | ||
BC147629.1 | - | 3 | 357 | 9056619 | ||
BC172110.1 | - | 3 | 329 | 9054396 |