Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034910.3 | NP_001030082.1 | 3 | 408 | ||
GenBank mRNA | AK079042.1 | - | 3 | 409 | ||
BC146022.1 | - | 2 | 264 | 40131325 | ||
BC146024.1 | - | 2 | 264 | 40131327 | ||
DQ012046.1 | - | 3 | 408 |