Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101038.1 | NP_001094508.1 | 4 | 751 | ||
GenBank mRNA | AY971517.1 | - | 4 | 751 |