Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081294.1 | NP_001074763.1 | 3 | 993 | ||
GenBank mRNA | BC147639.1 | - | 3 | 941 | 9056629 | |
BC147641.1 | - | 3 | 941 | 9056631 | ||
BC147701.1 | - | 3 | 941 | 9087460 | ||
BC147708.1 | - | 3 | 941 | 9087467 |