Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017348.2 | NP_059044.1 | 5, 6 | 1506 | ||
GenBank mRNA | BC168238.1 | - | 5, 6 | 1484 | 5598758 |