Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpcal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017357.1 | NP_059053.1 | 4 | 560 | ||
XM_006238829.2 | XP_006238891.1 | 5 | 855 | |||
GenBank mRNA | D13125.1 | - | 4 | 560 | ||
FQ212482.1 | - | 5 | 688 |