Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50578
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
7
783
NM_001034069.1
NP_001029241.1
7
783
NM_001034070.1
NP_001029242.1
7
783
NM_001034071.1
NP_001029243.1
7
783
NM_001034072.1
NP_001029244.1
7
783
NM_001034073.1
NP_001029245.1
6
720
NM_001034074.1
NP_001029246.1
6
720
NM_001034075.1
NP_001029247.1
6
720
NM_001301336.1
NP_001288265.1
7
783
NM_001301342.1
NP_001288271.1
7
783
NM_001301736.1
NP_001288665.1
6
720
NM_019131.2
NP_062004.1
6
720
XM_006243386.2
XP_006243448.1
7
775
XM_006243388.2
XP_006243450.1
7
775
XM_006243389.2
XP_006243451.1
7
775
XM_006243390.2
XP_006243452.1
7
775
XM_006243393.1
XP_006243455.1
6
718
XM_006243394.1
6
736
XM_006243395.2
XP_006243457.1
6
727
XM_008766207.1
XP_008764429.1
7
775
XM_008766208.1
XP_008764430.1
6
718
GenBank mRNA
AF372215.1
-
6
569
AF372216.1
-
7
682
AF372217.1
-
7
688
BC078780.1
-
6
687
7106337
CF108680.1
-
7
731
FQ214803.1
-
7
754
FQ214809.1
-
7
752
FQ214835.1
-
7
752
FQ214864.1
-
7
756
FQ214868.1
-
7
752
FQ214869.1
-
7
758
FQ214872.1
-
7
751
FQ214877.1
-
7
752
FQ214894.1
-
7
758
FQ214902.1
-
7
752
FQ214911.1
-
7
754
FQ214946.1
-
7
758
FQ214991.1
-
7
752
FQ215107.1
-
7
752
FQ215186.1
-
7
758
FQ215188.1
-
7
752
FQ215206.1
-
7
787
FQ215226.1
-
7
758
FQ215459.1
-
7
754
FQ215512.1
-
7
758
FQ215522.1
-
7
829
FQ215524.1
-
7
754
FQ215538.1
-
7
758
FQ215698.1
-
7
752
FQ215703.1
-
7
752
FQ215724.1
-
7
839
FQ215746.1
-
7
754
FQ215768.1
-
7
752
FQ215803.1
-
7
758
FQ215808.1
-
7
754
FQ215815.1
-
7
752
FQ215856.1
-
7
752
FQ215919.1
-
7
760
FQ215944.1
-
7
759
FQ215969.1
-
7
752
FQ216052.1
-
7
752
FQ216082.1
-
7
837
FQ216319.1
-
7
754
FQ216340.1
-
7
758
FQ216368.1
-
7
754
FQ216494.1
-
7
752
FQ216552.1
-
7
758
FQ216557.1
-
7
758
FQ216569.1
-
847
FQ216615.1
-
7
667
FQ216637.1
-
7
754
FQ216683.1
-
7
754
FQ216697.1
-
7
754
FQ217468.1
-
6
503
FQ217503.1
-
7
758
FQ217616.1
-
5
320
FQ217637.1
-
7
758
FQ217808.1
-
7
754
FQ223706.1
-
7
754
FQ223993.1
-
7
756
FQ224036.1
-
7
669
FQ224207.1
-
7
752
FQ224838.1
-
7
758
FQ224943.1
-
7
756
FQ224961.1
-
7
754
M34134.1
-
6
705
M34135.1
-
7
717
M34136.1
-
5
435
M60666.1
-
7
730
M60667.1
-
7
730
M60668.1
-
6
667
M60669.1
-
6
667
X02411.1
-
7
681
X02412.1
-
5
437
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0