Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177527.1 | NP_001170998.1 | 4 | 803 | ||
GenBank mRNA | BC138280.1 | - | 4 | 803 | 8861299 | |
BC145229.1 | - | 4 | 803 | 9053781 |