Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019152.2 | NP_062025.1 | 7 | 842 | ||
XM_006230706.2 | XP_006230768.1 | 7 | 901 | |||
XM_008760088.1 | XP_008758310.1 | 7 | 1473 | |||
XM_008760089.1 | XP_008758311.1 | 7 | 1153 | |||
GenBank mRNA | BC061880.1 | - | 7 | 842 | 5599301 | |
U53858.1 | - | 7 | 832 |