Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinlyp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037143.2 | NP_001032220.1 | 5 | 513 | ||
GenBank mRNA | AK009589.1 | - | 5 | 518 | ||
AK009604.1 | - | 5 | 515 | |||
AK009778.1 | - | 5 | 513 | |||
AK009924.1 | - | 5 | 515 | |||
BC139039.1 | - | 5 | 579 | 8862061 | ||
BC145686.1 | - | 5 | 579 | 40130989 |