Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022996.1 | NP_075372.1 | 5 | 601 | ||
GenBank mRNA | AF220209.1 | - | 5 | 639 | ||
AK050560.1 | - | 5 | 608 | |||
BC020359.1 | - | 5 | 588 | |||
BC026372.1 | - | 5 | 601 | 4162070 |