Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019156.2 | NP_062029.2 | 7 | 1176 | ||
GenBank mRNA | AY318963.1 | - | 1173 | |||
BC105821.1 | - | 7 | 1176 | 7371104 | ||
FQ209609.1 | - | 7 | 1189 | |||
FQ209682.1 | - | 7 | 1184 | |||
FQ209762.1 | - | 3 | 350 | |||
FQ210146.1 | - | 5 | 796 | |||
FQ210518.1 | - | 7 | 1181 | |||
FQ211325.1 | - | 3 | 310 | |||
FQ212575.1 | - | 7 | 1176 | |||
FQ218318.1 | - | 7 | 1188 | |||
FQ218401.1 | - | 5 | 749 | |||
FQ219297.1 | - | 7 | 1191 | |||
U44845.1 | - | 7 | 1166 |