Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039153.3 | NP_001034242.2 | 8 | 1082 | ||
GenBank mRNA | AK036568.1 | - | 8 | 1064 | ||
AK138661.1 | - | 8 | 1081 | |||
AK144772.1 | - | 8 | 1058 | |||
DQ278867.1 | - | 8 | 1083 |