Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cystm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081365.1 | NP_001074834.1 | 2 | 228 | ||
GenBank mRNA | AK002512.1 | - | 102 | |||
AK028217.1 | - | 99 | ||||
AK089711.1 | - | 2 | 90 | |||
AK167679.1 | - | 133 | ||||
BC028765.1 | - | 155 | 1195860 |